4-Methylisatin

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS24413G1 AS24413G5
ACTIAS24413G1EA 0 GBP
ACTIAS24413G1 ACTIAS24413G5
4-Methylisatin
4-Methylisatin
Formula: C₉H₇NO₂
MDL Number: MFCD00463404
CAS Number: 1128-44-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR