2-Amino-4,6-dichlorophenol

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3,5-Dichloro-6-hydroxyaniline

AS22177G25 AS22177G5
ACTIAS22177G25EA 0 GBP
ACTIAS22177G25 ACTIAS22177G5
2-Amino-4,6-dichlorophenol
2-Amino-4,6-dichlorophenol
Formula: C₆H₅Cl₂NO
MW: 178.01 g/mol
MDL Number: MFCD00035766
CAS Number: 527-62-8
EINECS: 208-421-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR