5-Chloro-2-(trifluoromethyl)benzamide
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
005709-1G
005709-5G
FLUO005709-1GEA
0
GBP
FLUO005709-1G
FLUO005709-5G
5-Chloro-2-(trifluoromethyl)benzamide
5-Chloro-2-(trifluoromethyl)benzamide
Formula:
C₈H₅ClF₃NO |
MDL Number:
MFCD01631406 CAS Number: 654-94-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...