6-Chloro-2-fluoro-3-methylbenzamide
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
005645-5G
005645-1G
FLUO005645-5GEA
0
GBP
FLUO005645-5G
FLUO005645-1G
6-Chloro-2-fluoro-3-methylbenzamide
6-Chloro-2-fluoro-3-methylbenzamide
Formula:
C₈H₇ClFNO |
MDL Number:
MFCD01631462 CAS Number: 286474-61-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...