2-Chloro-5-(trifluoromethyl)benzamide

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

005708-1G 005708-5G
FLUO005708-1GEA 0 GBP
FLUO005708-1G FLUO005708-5G
2-Chloro-5-(trifluoromethyl)benzamide
2-Chloro-5-(trifluoromethyl)benzamide
Formula: C₈H₅ClF₃NO
MDL Number: MFCD01631467
CAS Number: 20566-93-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR