4-Bromoisatin

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS21218G100 AS21218G25
ACTIAS21218G100EA 0 GBP
ACTIAS21218G100 ACTIAS21218G25
4-Bromoisatin
4-Bromoisatin
Formula: C₈H₄BrNO₂
MDL Number: MFCD03618555
CAS Number: 20780-72-7

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR