1-(2-Phenylethyl)piperazine
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
009824-5G
009824-25G
FLUO009824-5GEA
0
GBP
FLUO009824-5G
FLUO009824-25G
1-(2-Phenylethyl)piperazine
1-(2-Phenylethyl)piperazine
Formula:
C₁₂H₁₈N₂ |
MDL Number:
MFCD00040740 CAS Number: 5321-49-3 EINECS: 226-186-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...