Chlorodimethyloctadecylsilane, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5


M-Clarity™ quality level = MQ100

289108-25G 289108-5G
SIAL289108-25GEA 66.5 GBP
SIAL289108-25G SIAL289108-5G
Chlorodimethyloctadecylsilane, Sigma-Aldrich®
Chlorodimethyloctadecylsilane
Formula: C₂₀H₄₃ClSi
MW: 347.10 g/mol
MDL Number: MFCD00000504
CAS Number: 18643-08-8
EINECS: 242-472-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR