6-Bromo-2-chloro-3-methylphenol

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OT-1224-5G OT-1224-1G
COBBOT-1224-5GEA 0 GBP
COBBOT-1224-5G COBBOT-1224-1G
6-Bromo-2-chloro-3-methylphenol
6-Bromo-2-chloro-3-methylphenol
Formula: C₇H₆BrClO
MDL Number: MFCD16295099
CAS Number: 1226808-67-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR