3-Fluoro-4-(piperidine-1-carbonyl)phenylboronic acid

Supplier: COMBI-BLOCKS INC.
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

BB-3306-5G BB-3306-1G
COBBBB-3306-5GEA 0 GBP
COBBBB-3306-5G COBBBB-3306-1G
3-Fluoro-4-(piperidine-1-carbonyl)phenylboronic acid
3-Fluoro-4-(piperidine-1-carbonyl)phenylboronic acid
Formula: C₁₂H₁₅BFNO₃
MDL Number: MFCD08436039
CAS Number: 874289-10-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR