3-(Piperidin-1-ylmethyl)phenylboronic acid hydrochloride
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BB-5194-5G
BB-5194-1G
COBBBB-5194-5GEA
0
GBP
COBBBB-5194-5G
COBBBB-5194-1G
3-(Piperidin-1-ylmethyl)phenylboronic acid hydrochloride
3-(Piperidin-1-ylmethyl)phenylboronic acid hydrochloride
Formula:
C₁₂H₁₉BClNO₂ |
MDL Number:
MFCD10699627 CAS Number: 1072946-21-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...