4-(4-Methylpiperidine-1-carbonyl)phenylboronic acid
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BB-3023-1G
BB-3023-5G
COBBBB-3023-1GEA
0
GBP
COBBBB-3023-1G
COBBBB-3023-5G
4-(4-Methylpiperidine-1-carbonyl)phenylboronic acid
4-(4-Methylpiperidine-1-carbonyl)phenylboronic acid
Formula:
C₁₃H₁₈BNO₃ |
MDL Number:
MFCD04115676 CAS Number: 850568-21-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...