3-(Piperidin-1-ylmethyl)phenylboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

BB-5195-5G BB-5195-1G
COBBBB-5195-5GEA 0 GBP
COBBBB-5195-5G COBBBB-5195-1G
3-(Piperidin-1-ylmethyl)phenylboronic acid
3-(Piperidin-1-ylmethyl)phenylboronic acid
Formula: C₁₂H₁₈BNO₂
MDL Number: MFCD08060620
CAS Number: 1032610-59-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR