4-(3-(Ethoxycarbonyl)piperidine-1-carbonyl)phenylboronic acid pinacol ester

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Ethyl 1-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoyl)piperidine-3-carboxylate, 4-(3-Ethoxycarbonyl-1-piperidinylcarbonyl)benzeneboronic acid pinacol ester

PN-8657-5G PN-8657-1G
COBBPN-8657-5GEA 0 GBP
COBBPN-8657-5G COBBPN-8657-1G
4-(3-(Ethoxycarbonyl)piperidine-1-carbonyl)phenylboronic acid pinacol ester
4-(3-(Ethoxycarbonyl)piperidine-1-carbonyl)phenylboronic acid pinacol ester
Formula: C₂₁H₃₀BNO₅
MW: 387.28 g/mol
MDL Number: MFCD06657751
CAS Number: 850411-14-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR