4-(Piperidine-1-carbonyl)phenylboronic acid pinacol ester

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-(1-Piperidinylcarbonyl)benzeneboronic acid pinacol ester, Piperidin-1-yl(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)methanone

PN-3003-1G PN-3003-25G PN-3003-5G
COBBPN-3003-1GEA 0 GBP
COBBPN-3003-1G COBBPN-3003-25G COBBPN-3003-5G
4-(Piperidine-1-carbonyl)phenylboronic acid pinacol ester
4-(Piperidine-1-carbonyl)phenylboronic acid pinacol ester
Formula: C₁₈H₂₆BNO₃
MW: 315.21 g/mol
MDL Number: MFCD09027083
CAS Number: 938043-31-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR