3-(Piperidine-1-carbonyl)phenylboronic acid pinacol ester

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

PN-3053-5G PN-3053-1G
COBBPN-3053-5GEA 0 GBP
COBBPN-3053-5G COBBPN-3053-1G
3-(Piperidine-1-carbonyl)phenylboronic acid pinacol ester
3-(Piperidine-1-carbonyl)phenylboronic acid pinacol ester
Formula: C₁₈H₂₆BNO₃
MDL Number: MFCD09027084
CAS Number: 1073353-62-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR