1-(4-Bromophenyl)cyclobutanecarboxylic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HC-2989-5G HC-2989-1G
COBBHC-2989-5GEA 0 GBP
COBBHC-2989-5G COBBHC-2989-1G
1-(4-Bromophenyl)cyclobutanecarboxylic acid
1-(4-Bromophenyl)cyclobutanecarboxylic acid
Formula: C₁₁H₁₁BrO₂
MW: 255.1078 g/mol
MDL Number: MFCD08443165
CAS Number: 151157-49-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR