3-(2-(Piperidin-1-yl)ethylcarbamoyl)phenylboronic acid hydrochloride
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BB-3849-5G
BB-3849-1G
COBBBB-3849-5GEA
0
GBP
COBBBB-3849-5G
COBBBB-3849-1G
3-(2-(Piperidin-1-yl)ethylcarbamoyl)phenylboronic acid hydrochloride
3-(2-(Piperidin-1-yl)ethylcarbamoyl)phenylboronic acid hydrochloride
Formula:
C₁₄H₂₂BClN₂O₃ |
MDL Number:
MFCD11053816 CAS Number: 1072945-72-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...