4-Fluoro-3-morpholinobenzaldehyde

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS22468G1 AS22468G5
ACTIAS22468G1EA 0 GBP
ACTIAS22468G1 ACTIAS22468G5
4-Fluoro-3-morpholinobenzaldehyde
4-Fluoro-3-morpholinobenzaldehyde
Formula: C₁₁H₁₂FNO₂
MDL Number: MFCD13191604
CAS Number: 1197193-13-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR