2-Chloro-4-morpholinobenzaldehyde

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS21547G1 AS21547G25 AS21547G5
ACTIAS21547G1EA 76 GBP
ACTIAS21547G1 ACTIAS21547G25 ACTIAS21547G5
2-Chloro-4-morpholinobenzaldehyde
2-Chloro-4-morpholinobenzaldehyde
Formula: C₁₁H₁₂ClNO₂
MDL Number: MFCD06740104
CAS Number: 886501-36-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR