4-Morpholinobenzaldehyde

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS20626G25 AS20626G5
ACTIAS20626G25EA 125 GBP
ACTIAS20626G25 ACTIAS20626G5
4-Morpholinobenzaldehyde
4-Morpholinobenzaldehyde
Formula: C₁₁H₁₃NO₂
MW: 191.2264 g/mol
MDL Number: MFCD00735826
CAS Number: 1204-86-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR