3-Morpholinobenzaldehyde
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AS20585G1
AS20585G5
ACTIAS20585G1EA
0
GBP
ACTIAS20585G1
ACTIAS20585G5
3-Morpholinobenzaldehyde
3-Morpholinobenzaldehyde
Formula:
C₁₁H₁₃NO₂ |
MDL Number:
MFCD07772811 CAS Number: 446866-87-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...