4-(3-Bromophenyl)morpholine hydrochloride

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OT-0974-5G OT-0974-25G OT-0974-1G
COBBOT-0974-5GEA 0 GBP
COBBOT-0974-5G COBBOT-0974-25G COBBOT-0974-1G
4-(3-Bromophenyl)morpholine hydrochloride
4-(3-Bromophenyl)morpholine hydrochloride
Formula: C₁₀H₁₃BrClNO
MDL Number: MFCD12913975
CAS Number: 197846-83-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR