3-Bromo-4-morpholinobenzaldehyde
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AS21544G1
AS21544G25
AS21544G5
ACTIAS21544G1EA
0
GBP
ACTIAS21544G1
ACTIAS21544G25
ACTIAS21544G5
3-Bromo-4-morpholinobenzaldehyde
3-Bromo-4-morpholinobenzaldehyde
Formula:
C₁₁H₁₂BrNO₂ |
MDL Number:
MFCD06656549 CAS Number: 263349-24-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...