6-Bromo-4-quinazolone

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 6-Bromo-4(3H)-quinazolinone

HC-6043-5G HC-6043-1G
COBBHC-6043-5GEA 0 GBP
COBBHC-6043-5G COBBHC-6043-1G
6-Bromo-4-quinazolone
6-Bromo-4-quinazolone
Formula: C₈H₅BrN₂O
MW: 225.04 g/mol
MDL Number: MFCD01219541
CAS Number: 32084-59-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR