5-Cyano-2-thiopheneboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: (5-Cyanothiophen-2-yl)boronic acid, 5-Cyanothiophene-2-boronic acid

BB-8354-5G BB-8354-25G BB-8354-1G
COBBBB-8354-5GEA 0 GBP
COBBBB-8354-5G COBBBB-8354-25G COBBBB-8354-1G
5-Cyano-2-thiopheneboronic acid
5-Cyano-2-thiopheneboronic acid
Formula: C₅H₄BNO₂S
MW: 152.97 g/mol
MDL Number: MFCD02094029
CAS Number: 305832-67-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR