5-Nitro-3-picoline

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OT-0073-5G OT-0073-1G
COBBOT-0073-5GEA 0 GBP
COBBOT-0073-5G COBBOT-0073-1G
5-Nitro-3-picoline
5-Nitro-3-picoline
Formula: C₆H₆N₂O₂
MDL Number: MFCD04114214
CAS Number: 6960-20-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR