5-Nitro-2-picoline
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
OS-7442-1G
OS-7442-25G
OS-7442-5G
COBBOS-7442-1GEA
0
GBP
COBBOS-7442-1G
COBBOS-7442-25G
COBBOS-7442-5G
5-Nitro-2-picoline
5-Nitro-2-picoline
Formula:
C₆H₆N₂O₂ |
MDL Number:
MFCD04114179 CAS Number: 21203-68-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...