4-Nitro-2-picoline

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-Methyl-4-nitropyridine

AS21655G25 AS21655G5
ACTIAS21655G25EA 0 GBP
ACTIAS21655G25 ACTIAS21655G5
4-Nitro-2-picoline
4-Nitro-2-picoline
Formula: C₆H₆N₂O₂
MW: 138.1200 g/mol
MDL Number: MFCD03095077
CAS Number: 13508-96-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR