2-(Cyclopentylamino)-3-nitro-5-picoline

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

PY-7319-1G PY-7319-5G
COBBPY-7319-1GEA 0 GBP
COBBPY-7319-1G COBBPY-7319-5G
2-(Cyclopentylamino)-3-nitro-5-picoline
2-(Cyclopentylamino)-3-nitro-5-picoline
Formula: C₁₁H₁₅N₃O₂
MDL Number: MFCD10699704
CAS Number: 1033202-29-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR