2-Acetyl-5-methylpyridine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1-(5-Methylpyridin-2-yl)ethanone

AS21760G1 AS21760G5
ACTIAS21760G1EA 84 GBP
ACTIAS21760G1 ACTIAS21760G5
2-Acetyl-5-methylpyridine
2-Acetyl-5-methylpyridine
Formula: C₈H₉NO
MW: 135.1600 g/mol
MDL Number: MFCD06658292
CAS Number: 5308-63-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR