5-Acetyl-2-methoxypyridine
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SS-3001-1G
SS-3001-25G
SS-3001-5G
COBBSS-3001-1GEA
0
GBP
COBBSS-3001-1G
COBBSS-3001-25G
COBBSS-3001-5G
5-Acetyl-2-methoxypyridine
5-Acetyl-2-methoxypyridine
Formula:
C₈H₉NO₂ MW: 151.1600 g/mol |
MDL Number:
MFCD05664350 CAS Number: 213193-32-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...