3-Acetyl-2-chloropyridine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3-acetyl-2-chloropyridine, 1-(2-Chloro-3-pyridinyl)-1-ethanone, 1-(2-Chloropyridin-3-yl)ethanone

AS22609G1 AS22609G5
ACTIAS22609G1EA 0 GBP
ACTIAS22609G1 ACTIAS22609G5
3-Acetyl-2-chloropyridine
3-Acetyl-2-chloropyridine
Formula: C₇H₆ClNO
MW: 155.5816 g/mol
MDL Number: MFCD03840751
CAS Number: 55676-21-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR