2-Chloro-3-cyano-4-picoline

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-Chloro-4-methylpyridine-3-carbonitrile

AS20816G1 AS20816G5
ACTIAS20816G1EA 0 GBP
ACTIAS20816G1 ACTIAS20816G5
2-Chloro-3-cyano-4-picoline
2-Chloro-3-cyano-4-picoline
Formula: C₇H₅ClN₂
MW: 152.5810 g/mol
MDL Number: MFCD07082888
CAS Number: 65169-38-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR