5-Bromo-2-hydroxy-3-(trifluoromethyl)pyridine

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 5-Bromo-3-(trifluoromethyl)pyridin-2-ol

PY-7981-1G PY-7981-5G
COBBPY-7981-1GEA 0 GBP
COBBPY-7981-1G COBBPY-7981-5G
5-Bromo-2-hydroxy-3-(trifluoromethyl)pyridine
5-Bromo-2-hydroxy-3-(trifluoromethyl)pyridine
Formula: C₆H₃BrF₃NO
MW: 242.00 g/mol
MDL Number: MFCD12546486
CAS Number: 76041-79-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR