2-Acetyl-6-bromopyridine

Supplier: ACTIVATE SCIENTIFIC GMBH
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ACTIAS22067G1EA 0 GBP
ACTIAS22067G1 ACTIAS22067G25 ACTIAS22067G5
2-Acetyl-6-bromopyridine
2-Acetyl-6-bromopyridine
Formula: C₇H₆BrNO
MW: 200.03 g/mol
MDL Number: MFCD00272200
CAS Number: 49669-13-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR