2-Bromo-5-cyano-3-picoline
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AS31066G1
AS31066G5
ACTIAS31066G1EA
106
GBP
ACTIAS31066G1
ACTIAS31066G5
2-Bromo-5-cyano-3-picoline
2-Bromo-5-cyano-3-picoline
Formula:
C₇H₅BrN₂ MW: 197.0320 g/mol |
MDL Number:
MFCD03095063 CAS Number: 374633-37-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...