N-Methyl-4-(methylamino)picolinamide

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

PY-7788-5G PY-7788-1G
COBBPY-7788-5GEA 0 GBP
COBBPY-7788-5G COBBPY-7788-1G
N-Methyl-4-(methylamino)picolinamide
N-Methyl 4-(methylamino)picolinamide
Formula: C₈H₁₁N₃O
MDL Number: MFCD11504981
CAS Number: 1065074-98-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR