2-(Methylamino)nicotinonitrile

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

PY-7984-1G PY-7984-5G
COBBPY-7984-1GEA 0 GBP
COBBPY-7984-1G COBBPY-7984-5G
2-(Methylamino)nicotinonitrile
2-(Methylamino)nicotinonitrile
Formula: C₇H₇N₃
MW: 133.1506 g/mol
MDL Number: MFCD08689752
CAS Number: 52583-87-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR