3-Bromo-2-(methylthio)pyridine

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

PY-1424-1G PY-1424-5G
COBBPY-1424-1GEA 0 GBP
COBBPY-1424-1G COBBPY-1424-5G
3-Bromo-2-(methylthio)pyridine
3-Bromo-2-(methylthio)pyridine
Formula: C₆H₆BrNS
MDL Number: MFCD12964310
CAS Number: 51933-77-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR