Potassium-1-boc-6-chloroindole-2-trifluoroborate

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

TB-3548-1G TB-3548-5G
COBBTB-3548-1GEA 0 GBP
COBBTB-3548-1G COBBTB-3548-5G
Potassium-1-boc-6-chloroindole-2-trifluoroborate
Potassium 1-Boc-6-chloroindole-2-trifluoroborate
Formula: C₁₃H₁₃BClF₃KNO₂
MDL Number: MFCD11054034
CAS Number: 1073468-33-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR