Potassium(5-methyl-2-pyridyl)trifluoroborate

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

TB-2393-1G TB-2393-5G
COBBTB-2393-1GEA 0 GBP
COBBTB-2393-1G COBBTB-2393-5G
Potassium(5-methyl-2-pyridyl)trifluoroborate
Potassium (5-methyl-2-pyridyl)trifluoroborate
Formula: C₆H₆BF₃KN
MDL Number: MFCD09992972
CAS Number: 1073468-31-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR