Potassium(3-pyridyl)trifluoroborate
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Potassium trifluoro(pyridin-3-yl)borate
TB-2359-1G
TB-2359-5G
COBBTB-2359-1GEA
0
GBP
COBBTB-2359-1G
COBBTB-2359-5G
Potassium(3-pyridyl)trifluoroborate
Potassium (3-pyridyl)trifluoroborate
Formula:
C₅H₄BF₃KN MW: 185.0000 g/mol |
MDL Number:
MFCD09038545 CAS Number: 561328-69-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...