Potassium(2-pyridyl)trifluoroborate

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

TB-2096-1G TB-2096-5G
COBBTB-2096-1GEA 0 GBP
COBBTB-2096-1G COBBTB-2096-5G
Potassium(2-pyridyl)trifluoroborate
Potassium (2-pyridyl)trifluoroborate
Formula: C₅H₄BF₃KN
MDL Number: MFCD09992919
CAS Number: 561328-70-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR