4-(Boc-amino)cyclohexanone

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-Aminocyclohexan-1-one, N-BOC protected, tert-Butyl (4-oxocyclohexyl)carbamate

AM-1608-5G AM-1608-25G AM-1608-1G
COBBAM-1608-5GEA 0 GBP
COBBAM-1608-5G COBBAM-1608-25G COBBAM-1608-1G
4-(Boc-amino)cyclohexanone
4-(Boc-amino)cyclohexanone
Formula: C₁₁H₁₉NO₃
MW: 213.2700 g/mol
MDL Number: MFCD00798168
CAS Number: 179321-49-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR