trans-4-(Boc-amino)cyclohexanol

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

024189-1G 024189-5G
FLUO024189-1GEA 0 GBP
FLUO024189-1G FLUO024189-5G
trans-4-(Boc-amino)cyclohexanol
trans-4-(Boc-amino)cyclohexanol
Formula: C₁₁H₂₁NO₃
MDL Number: MFCD03844613
CAS Number: 111300-06-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR