trans-4-(Boc-amino)cyclohexanol
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
tert-Butyl (trans-4-hydroxycyclohexyl)carbamate, trans-4-Aminocyclohexan-1-ol, N-BOC protected
AM-1607-5G
AM-1607-25G
COBBAM-1607-5GEA
0
GBP
COBBAM-1607-5G
COBBAM-1607-25G
trans-4-(Boc-amino)cyclohexanol
trans-4-(Boc-amino)cyclohexanol
Formula:
C₁₁H₂₁NO₃ MW: 215.28 g/mol |
MDL Number:
MFCD03844613 CAS Number: 111300-06-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...