4-Amino-8-(trifluoromethyl)quinoline
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CFF814.1
005480-500MG
005480-1G
CHMPCFF814.1EA
0
GBP
CHMPCFF814.1
FLUO005480-500MG
FLUO005480-1G
4-Amino-8-(trifluoromethyl)quinoline
4-Amino-8-(trifluoromethyl)quinoline
Formula:
C₁₀H₇F₃N₂ |
MDL Number:
MFCD00269346 CAS Number: 243977-15-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...