4-Amino-7-(trifluoromethyl)quinoline

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

CFF813.1 005479-500MG 005479-1G
CHMPCFF813.1EA 0 GBP
CHMPCFF813.1 FLUO005479-500MG FLUO005479-1G
4-Amino-7-(trifluoromethyl)quinoline
4-Amino-7-(trifluoromethyl)quinoline
Formula: C₁₀H₇F₃N₂
MDL Number: MFCD00269411
CAS Number: 243666-11-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR