8-(Ethylamino)-5-nitroquinoline

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HC-2347-1G HC-2347-5G
COBBHC-2347-1GEA 0 GBP
COBBHC-2347-1G COBBHC-2347-5G
8-(Ethylamino)-5-nitroquinoline
8-(Ethylamino)-5-nitroquinoline
Formula: C₁₁H₁₁N₃O₂
MDL Number: MFCD00777818
CAS Number: 332402-92-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR